Invitrogen
Manufacturer Code:PA517627
Catalog # PIPA517627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to human and mouse RKIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 23 kDa morphine-binding protein; Basic cytosolic 21 kDa protein; epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNPpp; HCNPpp PEBP-1 Raf kinase inhibitory protein hippocampal cholinergic neurostimulating peptide neuropolypeptide h3 phosphatidylethanolamine-binding protein 1 prostatic binding protein prostatic-binding protein raf kinase inhibitor protein; Hippocampal cholinergic neurostimulating peptide; Neuropolypeptide h3; P23K; PEBP-1; Phosphatidylethanolamine-binding protein 1; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; Raf-1 inhibitor protein; Raf-1 kinase inhibitor protein; RKIP
Gene Aliases: HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; PBP; Pbp1; Pbpr; PEBP; PEBP-1; PEBP1; RKIP
UniProt ID: (Bovine) P13696, (Human) P30086, (Rat) P31044, (Mouse) P70296
Entrez Gene ID: (Bovine) 431786, (Human) 5037, (Rat) 29542, (Mouse) 23980
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.