Abnova
Manufacturer Code:PAB14533
Catalog # 89-117-124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified rat Pebp1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native Pebp1.
For Research Use Only
Protein Aliases: 23 kDa morphine-binding protein; HCNP; HCNPpp; Hippocampal cholinergic neurostimulating peptide; P23K; Pbp; PEBP-1; Phosphatidylethanolamine-binding protein 1; Raf-1 kinase inhibitor protein; Rkip
Gene Aliases: HCNP; Pbp; Pebp; Pebp1; Rkip
UniProt ID: (Rat) P31044
Entrez Gene ID: (Rat) 29542
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.