Invitrogen
Manufacturer Code:PA521057
Catalog # PIPA521057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the carboxy terminus of human PEAR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPEAR1; Jagged and Delta protein; Jedi; mPEAR1; Multiple EGF-like domains protein 12; multiple EGF-like-domains 12; Multiple epidermal growth factor-like domains protein 12; Platelet endothelial aggregation receptor 1; Protein Jedi; RP11-107D16.3 JEDI MEGF12 hPEAR1 multiple EGF-like domains protein 12 multiple EGF-like-domains 12 multiple epidermal growth factor-like domains protein 12
Gene Aliases: 3110045G13Rik; JEDI; Jedi-1; MEGF12; PEAR1; RGD1305653
UniProt ID: (Human) Q5VY43, (Mouse) Q8VHL7
Entrez Gene ID: (Human) 375033, (Mouse) 73182, (Rat) 295293
Molecular Function: extracellular matrix protein extracellular matrix structural protein membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.