Abnova
Manufacturer Code:H00005174M07A
Catalog # 89-105-083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E9 |
Immunogen | PDZK1 (AAH064961 a.a. &bsim 519 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant PDZK1.
For Research Use Only
Protein Aliases: CAP70; CFTR-associated protein of 70 kDa; CLAMP; Na(+)/H(+) exchange regulatory cofactor NHE-RF3; Na(+)/H(+) exchanger regulatory factor 3; Na/Pi cotransporter C-terminal-associated protein 1; NaPi-Cap1; NHERF-3; NHERF3; PDZ domain-containing protein 1; PDZ-containing kidney protein 1; PDZD1; Sodium-hydrogen exchanger regulatory factor 3
Gene Aliases: CAP70; CLAMP; NHERF-3; NHERF3; PDZD1; PDZK1
UniProt ID: (Human) Q5T2W1
Entrez Gene ID: (Human) 5174
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.