Proteintech
Manufacturer Code:105072AP150UL
Catalog # 50-172-6362
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDZK1 Fusion Protein Ag0787 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-terminal linking and modulating protein; C-terminal-linking and modulating protein; CAP70 CLAMP NaPi Cap1 NHERF 3 NHERF3 PDZ domain containing 1 PDZD1 PDZK1; CFTR-associated protein of 70 kDa; Dietary Pi-regulated RNA-1; Diphor-1; Na(+)/H(+) exchange regulatory cofactor NHE-RF3; Na(+)/H(+) exchanger regulatory factor 3; Na/Pi cotransporter C-terminal-associated protein 1; NaPi-Cap1; NHERF-3; PDZ domain-containing protein 1; PDZ-containing kidney protein 1; Sodium-hydrogen exchanger regulatory factor 3
Gene Aliases: 1700023D20Rik; 2610507N21Rik; 4921513F16Rik; AI267131; AI314638; AL022680; CAP70; CLAMP; D3Ertd537e; Diphor1; mPDZK1; NHERF-3; NHERF3; PDZD1; PDZK1
UniProt ID: (Human) Q5T2W1, (Mouse) Q9JIL4, (Rat) Q9JJ40
Entrez Gene ID: (Human) 5174, (Mouse) 59020, (Rat) 65144
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.