Proteintech
Manufacturer Code:187951AP150UL
Catalog # 50-173-2225
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDZD3 Fusion Protein Ag6931 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IKEPP NaPi Cap2 NHERF 4 NHERF4 PDZ domain containing 3 PDZD3 PDZK2; intestinal and kidney enriched PDZ protein; Intestinal and kidney-enriched PDZ protein; Na(+)/H(+) exchange regulatory cofactor NHE-RF4; Na/Pi cotransporter C-terminal-associated protein 2; naPi-Cap2; Natrium-phosphate cotransporter IIa C-terminal-associated protein 2; NHERF-4; PDZ domain containing 2; PDZ domain-containing protein 2; PDZ domain-containing protein 3; Sodium-hydrogen exchanger regulatory factor 4; sodium-phosphate cotransporter IIa C-terminal-associated protein 2
Gene Aliases: DLNB27; IKEPP; NaPi-Cap2; NHERF4; PDZD3; PDZK2; RGD1559807
UniProt ID: (Human) Q86UT5, (Mouse) Q99MJ6
Entrez Gene ID: (Human) 79849, (Mouse) 170761, (Rat) 500986
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.