Invitrogen
Manufacturer Code:PA544764
Catalog # PIPA544764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PDXP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: chronophilin; Chronophin; PLP phosphatase; PLP-phosophatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; pyridoxal phosphatase; Pyridoxal phosphate phosphatase; reg I binding protein I; reg I-binding protein 1; testicular secretory protein Li 36
Gene Aliases: 1600027H05Rik; AB041662; CIN; dJ37E16.5; PDXP; PLP; PLPP; Rbp1
UniProt ID: (Human) Q96GD0, (Bovine) Q3ZBF9, (Mouse) P60487
Entrez Gene ID: (Human) 57026, (Bovine) 506308, (Dog) 100688635, (Mouse) 57028, (Rat) 727679
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.