Novus Biologicals
Manufacturer Code:NBP227461
Catalog # NBP227461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-QHDLVPHYYVES corresponding to C-Terminus according to NP_064711.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chronophin; chronophin CIN dJ37E16.5 EC 3.1.3 EC 3.1.3.74 FLJ32703 PLP PLP phosphatase PLPP pyridoxal (pyridoxine vitamin B6) phosphatase pyridoxal phosphate phosphatase; PLP phosphatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; Pyridoxal phosphate phosphatase; testicular secretory protein Li 36
Gene Aliases: CIN; dJ37E16.5; PDXP; PLP; PLPP
UniProt ID: (Human) Q96GD0
Entrez Gene ID: (Human) 57026
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.