Abnova
Manufacturer Code:PAB27578
Catalog # 89-129-089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human PDXP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of PDXP.
For Research Use Only
Protein Aliases: Chronophin; CIN; dJ37E16.5; FLJ32703; PLP; PLP phosphatase; pyridoxal (pyridoxine, vitamin B6) phosphatase; Pyridoxal phosphate phosphatase; testicular secretory protein Li 36
Gene Aliases: CIN; dJ37E16.5; PDXP; PLP; PLPP
UniProt ID: (Human) Q96GD0
Entrez Gene ID: (Human) 57026
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.