Invitrogen
Manufacturer Code:PA513842
Catalog # PIPA513842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PDXK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory sperm binding protein Li 1a; Pyridoxal kinase; Pyridoxal kinase Pyridoxine kinase PKH PNK; pyridoxamine kinase; Pyridoxine kinase; vitamin B6 kinase
Gene Aliases: 2310036D04Rik; AA119688; C21orf124; C21orf97; C77999; HEL-S-1a; PDXK; PKH; PNK; PRED79
UniProt ID: (Human) O00764, (Mouse) Q8K183
Entrez Gene ID: (Human) 8566, (Mouse) 216134
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.