Invitrogen
Manufacturer Code:PA590084
Catalog # PIPA590084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 489-788 of human PDXDC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: pyridoxal-dependent decarboxylase domain containing 1; Pyridoxal-dependent decarboxylase domain-containing protein 1
Gene Aliases: 2210010A19Rik; AA415817; KIAA0251; Kiaa0251-hp; LP8165; PDXDC1; RGD1562597
UniProt ID: (Human) Q6P996, (Mouse) Q99K01
Entrez Gene ID: (Human) 23042, (Mouse) 94184, (Rat) 304721
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.