Invitrogen
Manufacturer Code:PA572207
Catalog # PIPA572207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 136-169 amino acids from the Central region of Mouse Pdx1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; IPF-1; Islet/duodenum homeobox 1; Pancreas/duodenum homeobox protein 1; Pancreas/duodenum homeobox protein 1 PDX-1 Insulin promoter factor 1 IPF-1 Islet/duodenum homeobox-1 IDX-1 Somatostatin-transactivating factor 1 STF-1 Insulin upstream factor 1 IUF-1 Glucose-sensitive factor GSF PDX1 IPF1; pancreatic and duodenal homeobox gene 1; Somatostatin-transactivating factor 1; STF-1
Gene Aliases: IDX-1; IPF-1; Ipf1; Mody4; pdx-1; Pdx1; STF-1
UniProt ID: (Mouse) P52946
Entrez Gene ID: (Mouse) 18609
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.