Novus Biologicals
Manufacturer Code:NBP201512
Catalog # NBP201512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D12 |
Immunogen | Full length human recombinant protein of human PDSS2(NP_065114) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS2; All-trans-decaprenyl-diphosphate synthase subunit 2; All-trans-decaprenyl-diphosphate synthase subunit 2 bA59I9.3 C6orf210decaprenyl-diphosphate synthase subunit 2 Candidate tumor suppressor protein chromosome 6 open reading frame 210 Decaprenyl pyrophosphate synthase subunit 2 decaprenyl pyrophosphate synthetase subunit 2 DLP1hDLP1 EC 2.5.1.91 prenyl (decaprenyl) diphosphate synthase subunit 2 subunit 2 of decaprenyl diphosphate synthase; Candidate tumor suppressor protein; Decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; Decaprenyl-diphosphate synthase subunit 2; Solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase
Gene Aliases: bA59I9.3; C6orf210; COQ10D3; DLP1; hDLP1; PDSS2
UniProt ID: (Human) Q86YH6
Entrez Gene ID: (Human) 57107
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.