Invitrogen
Manufacturer Code:PA556230
Catalog # PIPA556230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PDSS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS2; All-trans-decaprenyl-diphosphate synthase subunit 2; Candidate tumor suppressor protein; Decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; Decaprenyl-diphosphate synthase subunit 2; prenyl (solanesyl) diphosphate synthase, subunit 2; prenyltransferase-like mitochondrial protein; RP11-59I9.3 C6orf210 DLP1 bA59I9.3 hDLP1 all-trans-decaprenyl-diphosphate synthase subunit 2 decaprenyl pyrophosphate synthase subunit 2 decaprenyl pyrophosphate synthetase subunit 2 decaprenyl-diphosphate synthase subunit 2 subunit 2 of decaprenyl diphosphate synthase; Solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase
Gene Aliases: 5430420P03Rik; bA59I9.3; C6orf210; COQ10D3; DLP1; Gm60; hDLP1; kd; mDLP1; PDSS2; Plmp
UniProt ID: (Human) Q86YH6, (Rat) Q5U2R1, (Mouse) Q33DR3
Entrez Gene ID: (Human) 57107, (Rat) 365592, (Mouse) 71365
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.