Invitrogen
Manufacturer Code:PA542831
Catalog # PIPA542831
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PDSS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS1; All-trans-decaprenyl-diphosphate synthase subunit 1; COenzyme Q (ubiquinone) biosynthesis family member (coq-1)-like; coenzyme Q1 homolog; decaprenyl diphdsphate synthase subunit1; Decaprenyl pyrophosphate synthase subunit 1; decaprenyl pyrophosphate synthetase subunit 1; Decaprenyl-diphosphate synthase subunit 1; polyprenyl pyrophosphate synthetase; Prenyl (decaprenyl) diphosphate synthase subunit 1 COQ1 MGC70953 RP13-16H11.3 TPRT TPT hDPS1 DPS SPS TPT 1; prenyl (decaprenyl) diphosphate synthase, subunit 1; prenyl diphosphate synthase, subunit 1; Solanesyl-diphosphate synthase subunit 1; subunit 1 of decaprenyl diphosphate synthase; subunit 1 of solanesyl diphosphate synthase; TPT 1; trans-prenyltransferase; trans-prenyltransferase (TPT); Trans-prenyltransferase 1
Gene Aliases: 2610203G20Rik; 2700031G06Rik; COQ1; COQ10D2; DPS; DPS1; hDPS1; mDLP1; mSPS1; PDSS1; SPS; Sps1; TPRT; TPT; TPT 1; wu:fc11f12; wu:fd05d05; zgc:112058
UniProt ID: (Human) Q5T2R2, (Mouse) Q33DR2
Entrez Gene ID: (Bovine) 509696, (Pig) 100514361, (Horse) 100065489, (Dog) 607944, (Rabbit) 100350927, (Human) 23590, (Guinea pig) 100721670, (Mouse) 56075, (Rat) 100364990, (Zebrafish) 550349
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.