Abnova
Manufacturer Code:PAB23207
Catalog # 89-124-908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human PDSS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant PDSS1.
For Research Use Only
Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS1; All-trans-decaprenyl-diphosphate synthase subunit 1; coenzyme Q1 homolog; COQ1; Decaprenyl pyrophosphate synthase subunit 1; Decaprenyl-diphosphate synthase subunit 1; DPS; hDPS1; MGC70953; polyprenyl pyrophosphate synthetase; RP13-16H11.3; Solanesyl-diphosphate synthase subunit 1; SPS; subunit 1 of decaprenyl diphosphate synthase; TPRT; TPT; TPT 1; trans-prenyltransferase (TPT); Trans-prenyltransferase 1
Gene Aliases: COQ1; COQ10D2; DPS; DPS1; hDPS1; PDSS1; SPS; TPRT; TPT; TPT 1
UniProt ID: (Human) Q5T2R2
Entrez Gene ID: (Human) 23590
Molecular Function: acyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.