Abnova
Manufacturer Code:H00023590K
Catalog # 89-201-239
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PDSS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PDSS1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS1; All-trans-decaprenyl-diphosphate synthase subunit 1; coenzyme Q1 homolog; COQ1 DPS MGC70953 RP13-16H11.3 SPS TPRT TPT hDPS1; Decaprenyl pyrophosphate synthase subunit 1; Decaprenyl-diphosphate synthase subunit 1; polyprenyl pyrophosphate synthetase; Solanesyl-diphosphate synthase subunit 1; subunit 1 of decaprenyl diphosphate synthase; TPT 1; trans-prenyltransferase (TPT); Trans-prenyltransferase 1
Gene Aliases: COQ1; COQ10D2; DPS; DPS1; hDPS1; PDSS1; SPS; TPRT; TPT; TPT 1
UniProt ID: (Human) Q5T2R2
Entrez Gene ID: (Human) 23590
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.