Invitrogen
Manufacturer Code:PA5100680
Catalog # PIPA5100680
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDP2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [pyruvate dehydrogenase (acetyl-transferring)]-phosphatase; [pyruvate dehydrogenase (acetyl-transferring)]-phosphatase [Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 2 mitochondrial 4833426J09Rik Gm1705 KIAA1348 mKIAA1348 PDP 2 Pdp2 PDPC 2 PPM2B PPM2C2 protein phosphatase 2C magnesium-dependent catalytic subunit 2 protein phosphatase Mg2+/Mn2+ dependent 2B pyruvate dehydrogenase [acetyl-transferring]-phosphatase 2 mitochondrial pyruvate dehydrogenase phosphatase catalytic subunit 2 pyruvate dehydrogenase phosphatase isoenzyme 2 pyruvate dehydrogenase phosphatase catalytic subunit 2 pyruvate dehyrogenase phosphatase catalytic subunit 2; [Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 2, mitochondrial; PDP 2; PDPC 2; protein phosphatase 2C, magnesium-dependent, catalytic subunit 2; protein phosphatase, Mg2+/Mn2+ dependent 2B; pyruvate dehydrogenase [acetyl-transferring]-phosphatase 2, mitochondrial; Pyruvate dehydrogenase phosphatase catalytic subunit 2; pyruvate dehydrogenase phosphatase isoenzyme 2; pyruvate dehydrogenase phosphatase, catalytic subunit 2
Gene Aliases: 4833426J09Rik; Gm1705; KIAA1348; mKIAA1348; PDP2; PPM2B; PPM2C2
UniProt ID: (Human) Q9P2J9, (Rat) O88484
Entrez Gene ID: (Human) 57546, (Rat) 246311, (Mouse) 382051
Molecular Function:
enzyme modulator
hydrolase
kinase inhibitor
kinase modulator
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.