Invitrogen
Manufacturer Code:PA578679
Catalog # PIPA578679
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PDP 1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial; FLJ32517 FLJ56179 MGC119646 PDH PDP PDPC PPM2C PDP1 PDP 1 "protein phosphatase 2C magnesium-dependent catalytic subunit " pyruvate dehydrogenase phosphatase catalytic subunit 1 pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase PDPC 1 "&lsqbPyruvate dehydrogenase &lsqbacetyl-transferring]]-phosphatase 1 mitochondrial " pyruvate dehyrogenase phosphatase catalytic subunit 1; PDP 1; PDPC 1; Protein phosphatase 2C; protein phosphatase 2C, magnesium dependent, catalytic subunit; protein phosphatase 2C, magnesium-dependent, catalytic subunit; protein phosphatase, Mg2+/Mn2+ dependent 2A; pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase; Pyruvate dehydrogenase phosphatase catalytic subunit 1; pyruvate dehydrogenase phosphatase isoenzyme 2
Gene Aliases: Gm1024; PDH; PDP; PDP1; PDPC; PPM2A; PPM2C
UniProt ID: (Human) Q9P0J1, (Mouse) Q3UV70
Entrez Gene ID: (Human) 54704, (Mouse) 381511
Molecular Function:
enzyme modulator
hydrolase
kinase inhibitor
kinase modulator
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.