Invitrogen
Manufacturer Code:PA545204
Catalog # PIPA545204
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PDLIM7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110003B01Rik; ENIGMA; enigma (LIM domain protein); enigma protein; LIM domain protein; LIM mineralization protein; LIM mineralization protein 2; Lim mineralization protein 3; LMP; PDZ and LIM domain 7 (enigma); PDZ and LIM domain protein 7; Protein enigma
Gene Aliases: ENIGMA; Lim1; LMP; LMP1; LMP3; PDLIM7; zgc:65907; zgc:85607
UniProt ID: (Human) Q9NR12, (Bovine) Q3SX40, (Mouse) Q3TJD7, (Rat) Q9Z1Z9, (Zebrafish) Q6P7E4
Entrez Gene ID: (Horse) 100058356, (Rabbit) 100357898, (Human) 9260, (Bovine) 533851, (Dog) 611246, (Guinea pig) 100712754, (Mouse) 67399, (Rat) 286908, (Zebrafish) 393813
Molecular Function: actin family cytoskeletal protein cytoskeletal protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.