Invitrogen
Manufacturer Code:PA5110094
Catalog # PIPA5110094
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 90-190 of human PDLIM7 (NP_998801.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110003B01Rik; BOS_7901 Enigma LIM mineralization protein Lim1 LMP LMP1 LMP3 pdlim7 PDZ and LIM domain protein 7 Protein enigma zgc:65907 zgc:85607; LIM domain protein; LIM mineralization protein; Lim mineralization protein 3; LMP; PDZ and LIM domain 7 (enigma); PDZ and LIM domain protein 7; Protein enigma
Gene Aliases: ENIGMA; LMP; LMP1; LMP3; PDLIM7
UniProt ID: (Human) Q9NR12, (Mouse) Q3TJD7
Entrez Gene ID: (Human) 9260, (Mouse) 67399
Molecular Function: actin family cytoskeletal protein cytoskeletal protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.