OriGene
Manufacturer Code:TA502711
Catalog # 50-167-3745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI9B6 |
Immunogen | Full length human recombit protein of human PDLIM2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MYSTIQUE SLIM; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique
Gene Aliases: MYSTIQUE; PDLIM2; PP6345; SLIM
UniProt ID: (Human) Q96JY6
Entrez Gene ID: (Human) 64236, (Dog) 486123
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.