Novus Biologicals
Manufacturer Code:NBP200619
Catalog # NBP200619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11A12 |
Immunogen | Full length human recombinant protein of human PDLIM2 (NP_067643) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ34715 MYSTIQUE PDZ and LIM domain 2 (mystique) PDZ and LIM domain protein 2 PDZ-LIM protein mystique SLIM; PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique
Gene Aliases: MYSTIQUE; PDLIM2; PP6345; SLIM
UniProt ID: (Human) Q96JY6
Entrez Gene ID: (Human) 64236
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.