Invitrogen
Manufacturer Code:PA513775
Catalog # PIPA513775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PDK3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; Pyruvate dehydrogenase &lsqblipoamide]] kinase isozyme 3 mitochondrial Pyruvate dehydrogenase kinase isoform 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Aliases: CMTX6; GS1-358P8.4; PDHK3; PDK3
UniProt ID: (Human) Q15120
Entrez Gene ID: (Human) 5165
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.