Proteintech
Manufacturer Code:122151AP150UL
Catalog # 50-172-7801
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDK3 Fusion Protein Ag2879 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; PDK3; pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Aliases: 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3
UniProt ID: (Human) Q15120, (Mouse) Q922H2
Entrez Gene ID: (Human) 5165, (Mouse) 236900
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.