Invitrogen
Manufacturer Code:MA535568
Catalog # PIMA535568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1113 |
Immunogen | A synthesized peptide derived from human PDK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2 mitochondrial BOS_18972 PDH kinase 2 PDHK2 PDK P45 Pdk2 PDKII Pyruvate dehydrogenase kinase isoform 2; [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 2, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 2, mitochondrial; PDH kinase 2; PDK P45; pyruvate dehydrogenase 2; pyruvate dehydrogenase kinase 2 subunit p45 (PDK2); Pyruvate dehydrogenase kinase isoform 2; pyruvate dehydrogenase kinase, isoenzyme 2; pyruvate dehydrogenase kinase, isozyme 2
Gene Aliases: Pdk2
UniProt ID: (Rat) Q64536, (Mouse) Q9JK42
Entrez Gene ID: (Rat) 81530, (Mouse) 18604
Molecular Function: kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.