Proteintech
Manufacturer Code:100261AP150UL
Catalog # 50-172-5979
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDK1 Fusion Protein Ag0025 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial; mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1; PDH kinase 1; PDH kinase 1 PDHK1 PDK1 Pyruvate dehydrogenase kinase isoform 1; Pyruvate dehydrogenase kinase isoform 1; pyruvate dehydrogenase kinase, isoenzyme 1; pyruvate dehydrogenase kinase, isozyme 1
Gene Aliases: B830012B01; D530020C15Rik; PDHK1; PDK1
UniProt ID: (Human) Q15118, (Mouse) Q8BFP9
Entrez Gene ID: (Human) 5163, (Mouse) 228026
Molecular Function:
kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.