R&D Systems
Manufacturer Code:AF864SP
Catalog # AF864SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH-coupled human PDK-1 synthetic peptide CKIIKLEYDFPEKFFPKARD Accession # AAD38505 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-phosphoinositide dependent protein kinase-1; 3-phosphoinositide-dependent protein kinase 1; 3-phosphoinositide-dependent protein kinase 1 3-phosphoinositide-dependent protein kinase 2 pseudogene PDK1 PDPK1 PDPK2 PDPK2P PKB kinase PkB kinase like gene 1; 3-phosphoinositide-dependent protein kinase 2 pseudogene; hPDK1; PkB kinase like gene 1
Gene Aliases: PDK1; PDPK1; PDPK2; PDPK2P; PRO0461
UniProt ID: (Human) O15530
Entrez Gene ID: (Human) 5170
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.