Invitrogen
Manufacturer Code:MA110032
Catalog # 01-670-289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 38H8 |
Immunogen | Yeast nuclear preparation. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CaO19.12595 protein disulfide isomerase PDI1 PDI1P Eps1p; PDI; protein disulfide isomerase PDI1; Protein disulfide-isomerase; Thioredoxin-related glycoprotein 1
Gene Aliases: MFP1; PDI1; TRG1; YCL043C; YCL313; YCL43C
UniProt ID: (Baker's yeast) P17967
Entrez Gene ID: (Baker's yeast) 850314
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.