Invitrogen
Manufacturer Code:MA529737
Catalog # PIMA529737
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 23 |
Immunogen | Recombit Mouse P4HB protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: CaO19.12595 protein disulfide isomerase PDI1 PDI1P Eps1p; Cellular thyroid hormone-binding protein; Endoplasmic reticulum resident protein 59; ER protein 59; p55; PDI; Prolyl 4-hydroxylase subunit beta; protein disulfide isomerase; Protein disulfide-isomerase
Gene Aliases: ERp59; P4hb; PDI; Pdia1; Thbp
UniProt ID: (Mouse) P09103
Entrez Gene ID: (Mouse) 18453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.