Invitrogen
Manufacturer Code:PA578098
Catalog # PIPA578098
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PDI. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cellular thyroid hormone-binding protein; collagen prolyl 4-hydroxylase beta; DSI ERBA2L GIT P4Hbeta PDI PDIA1 PHDB PO4DB PO4HB PROHB P4HB cellular thyroid hormone-binding protein protein disulfide-isomerase "protein disulfide isomerase family A member 1 " glutathione-insulin transhydrogenase protein disulfide isomerase/oxidoreductase "procollagen-proline 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase) beta polypeptide " collagen prolyl 4-hydroxylase beta protocollagen hydroxylase prolyl 4-hydroxylase subunit beta thyroid hormone-binding protein p55 protein disulfide isomerase-associated 1 p55 "prolyl 4-hydroxylase beta polypeptide " Endoplasmic reticulum resident protein 59; ER protein 59; glutathione-insulin transhydrogenase; p55; PDI; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase, beta polypeptide; protein disulfide isomerase; protein disulfide isomerase family A, member 1; protein disulfide isomerase-associated 1; protein disulfide isomerase/oxidoreductase; Protein disulfide-isomerase; protocollagen hydroxylase; testicular secretory protein Li 32; thyroid hormone-binding protein p55
Gene Aliases: CLCRP1; DSI; ERBA2L; ERp59; GIT; P4HB; P4Hbeta; PDI; PDIA1; PHDB; PO4DB; PO4HB; PROHB; Thbp
UniProt ID: (Human) P07237, (Mouse) P09103
Entrez Gene ID: (Human) 5034, (Mouse) 18453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.