Invitrogen
Manufacturer Code:PA577821
Catalog # PIPA577821
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Rat PDI synthetic peptide (409-509 aa C-terminal) conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2 PDI PDIA2 PDIP pancreatic protein disulfide isomerase; pdia2; PDIp; protein disulfide isomerase (pancreas) like; protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; Protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma
Gene Aliases: 1810041F13Rik; AI661267; PDA2; PDI; PDIA2; PDIP; Pdipl; PDIR; XELAEV_18045582mg; XPDIp
UniProt ID: (Human) Q6B010, (Mouse) D3Z6P0
Entrez Gene ID: (Rhesus monkey) 694964, (Bovine) 504356, (Human) 64714, (Sheep) 101122403, (Pig) 100738360, (Rat) 287164, (Mouse) 69191, (Guinea pig) 100713422, (Xenopus) 399040
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.