Novus Biologicals
Manufacturer Code:NBP299080F
Catalog # NB057841
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PDI. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pancreas-specific protein disulfide isomerase; Pancreas-specific protein disulfide isomerase pancreatic protein disulfide isomerase PDA2 PDI PDIp PDIPEC 5.3.4.1 PDIR protein disulfide isomerase A2 protein disulfide isomerase family A member 2 protein disulfide isomerase pancreatic protein disulfide isomerase-associated 2 protein disulfide-isomerase A2 Rho GDP dissociation inhibitor gamma; pancreatic protein disulfide isomerase; PDIp; protein disulfide isomerase A2; protein disulfide isomerase family A, member 2; protein disulfide isomerase, pancreatic; protein disulfide isomerase-associated 2; Protein disulfide-isomerase A2; Rho GDP dissociation inhibitor gamma
Gene Aliases: PDA2; PDI; PDIA2; PDIP; PDIR
UniProt ID: (Human) Q6B010
Entrez Gene ID: (Human) 64714
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.