Invitrogen
Manufacturer Code:MA535551
Catalog # PIMA535551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1074 |
Immunogen | A synthesized peptide derived from human Pyruvate Dehydrogenase E1 beta subunit |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610103L06Rik AL024199 C81408 Pdhb PDHBD PDHE1-B PHE1B Pyruvate dehydrogenase E1 component subunit beta mitochondrial; PDHE1-B; Pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase, E1 beta polypeptide
Gene Aliases: 2610103L06Rik; AL024199; C81408; PDHB; PDHBD; PDHE1-B; PHE1B
UniProt ID: (Human) P11177, (Rat) P49432, (Mouse) Q9D051
Entrez Gene ID: (Human) 5162, (Rat) 289950, (Mouse) 68263
Molecular Function: dehydrogenase lyase oxidoreductase transferase transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.