Proteintech
Manufacturer Code:147441AP150UL
Catalog # 50-172-9690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDHB Fusion Protein Ag6442 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDHB PDHE 1B PDHE1 B PDHE1B PHE1B Pyruvate Dehydrogenase E1 beta subunit; PDHE1-B; Pyruvate dehydrogenase E1 component subunit beta, mitochondrial; pyruvate dehydrogenase, E1 beta polypeptide
Gene Aliases: 2610103L06Rik; AL024199; C81408; PDHB; PDHBD; PDHE1-B; PHE1B
UniProt ID: (Human) P11177, (Rat) P49432, (Mouse) Q9D051
Entrez Gene ID: (Human) 5162, (Rat) 289950, (Mouse) 68263
Molecular Function:
dehydrogenase
lyase
oxidoreductase
transferase
transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.