Invitrogen
Manufacturer Code:PA598041
Catalog # PIPA598041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Pyruvate dehydrogenase E1 component subunit alpha testis-specific form mitochondrial protein (119-388AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDHE1-A type II; PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT, TESTIS-SPECIFIC FORM, MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II); Pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific
Gene Aliases: Pdha-2; PDHA2; PDHAL
UniProt ID: (Human) P29803, (Mouse) P35487
Entrez Gene ID: (Human) 5161, (Mouse) 18598
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.