Invitrogen
Manufacturer Code:PA5106454
Catalog # PIPA5106454
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDHA2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pdha2 Pdha-2 PDHAL PDHE1-A type II pyruvate dehydrogenase pyruvate dehydrogenase (lipoamide) alpha 2 pyruvate dehydrogenase E1 alpha 2 PYRUVATE DEHYDROGENASE E1 COMPONENT ALPHA SUBUNIT TESTIS-SPECIFIC FORM MITOCHONDRIAL PRECURSOR (PDHE1-A TYPE II) pyruvate dehydrogenase E1 component subunit alpha testis-specific form mitochondrial pyruvate dehydrogenase E1-alpha polypeptide testis specific; PDHE1-A type II; pyruvate dehydrogenase E1 alpha 2; Pyruvate dehydrogenase E1 component subunit alpha, testis-specific form, mitochondrial; pyruvate dehydrogenase, E1-alpha polypeptide, testis specific
Gene Aliases: PDHA2; PDHAL
UniProt ID: (Human) P29803, (Rat) Q06437
Entrez Gene ID: (Human) 5161, (Rat) 117098
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.