Invitrogen
Manufacturer Code:PA572684
Catalog # PIPA572684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 233&bsim263 amino acids from the Central region of Zebrafish PDHA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDHE1-A type I Pyruvate dehydrogenase E1 component subunit alpha somatic form mitochondrial PHE1A PDHA1pdha1apdha1b; pyruvate dehydrogenase (lipoamide) alpha 1; pyruvate dehydrogenase (lipoamide) alpha 1a; pyruvate dehydrogenase E1 alpha 1 subunit a
Gene Aliases: im:6895726; pdha1; wu:fd18b01; wu:fo96f03; zgc:73271; zgc:86692
Entrez Gene ID: (Zebrafish) 406702
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.