Invitrogen
Manufacturer Code:MA532950
Catalog # PIMA532950
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C4 |
Immunogen | Synthetic peptide corresponding to the C-terminus of Human Pyruvate Dehydrogenase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PDHE1-A type I; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; Pyruvate dehydrogenase E1 component subunit alpha somatic form mitochondrial PDHE1-A type I PDHA1 PHE1A; Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1alpha subunit
Gene Aliases: PDHA; Pdha-1; PDHA1; PDHAD; PDHCE1A; PHE1A
UniProt ID: (Human) P08559, (Mouse) P35486
Entrez Gene ID: (Human) 5160, (Mouse) 18597
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.