Invitrogen
Manufacturer Code:PA5101800
Catalog # PIPA5101800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDHA1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDHA PDHA1 Pdha-1 PDHAD PDHCE1A PDHE1-A type I PHE1A pyruvate dehydrogenase pyruvate dehydrogenase (lipoamide) alpha 1 pyruvate dehydrogenase alpha 1 pyruvate dehydrogenase complex E1-alpha polypeptide 1 pyruvate dehydrogenase E1 alpha 1 pyruvate dehydrogenase E1 alpha 1 subunit pyruvate dehydrogenase E1 component alpha subunit pyruvate dehydrogenase E1 component alpha subunit precursor pyruvate dehydrogenase E1 component subunit alpha somatic form mitochondrial pyruvate dehydrogenase E1 subunit pyruvate dehydrogenase E1 subunit alpha 1 pyruvate dehydrogenase E1alpha subunit; PDHE1-A type I; pyruvate dehydrogenase complex, E1-alpha polypeptide 1; pyruvate dehydrogenase E1 alpha 1; Pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial; pyruvate dehydrogenase E1 subunit; pyruvate dehydrogenase E1alpha subunit
Gene Aliases: PDHA; Pdha-1; PDHA1; PDHAD; PDHCE1A; PHE1A
UniProt ID: (Human) P08559, (Mouse) P35486
Entrez Gene ID: (Human) 5160, (Rat) 29554, (Mouse) 18597
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.