Novus Biologicals
Manufacturer Code:NBP143349JF646
Catalog # NBP43349646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | APB5 |
Immunogen | The immunogen for this antibody was PDGF receptor beta. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor CD140 antigen-like family member B CD140b CD140b antigen EC 2.7.10 EC 2.7.10.1 JTK12 PDGFR PDGFR1 PDGF-R-beta platelet-derived growth factor receptor beta polypeptide; CD140 antigen-like family member B; CD140b; PDGF-R-beta; PDGFR-1; PDGFR-beta; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor, beta polypeptide
Gene Aliases: CD140B; IBGC4; IMF1; JTK12; KOGS; PDGFR; PDGFR-1; PDGFR1; PDGFRB; PENTT
UniProt ID: (Human) P09619
Entrez Gene ID: (Human) 5159
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.