Invitrogen
Manufacturer Code:PA572002
Catalog # PIPA572002
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 16-44 amino acids from the N-terminal region of human PDGFRL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDGF receptor beta-like tumor suppressor; PDGFR-like protein; Platelet-derived growth factor receptor-like protein; Platelet-derived growth factor receptor-like protein PDGFR-like protein PDGF receptor beta-like tumor suppressor PDGFRL PRLTS; platelet-derived growth factor-beta-like tumor suppressor
Gene Aliases: 1110039P19Rik; AV013190; PDGFRL; PDGRL; PRLTS
UniProt ID: (Human) Q15198, (Mouse) Q6PE55
Entrez Gene ID: (Human) 5157, (Mouse) 68797
Molecular Function: kinase protein kinase protein kinase receptor receptor transferase tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.