Invitrogen
Manufacturer Code:PA5106500
Catalog # PIPA5106500
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDGFRL corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110039P19Rik AV013190 PDGF receptor beta-like tumor suppressor PDGFRL PDGFR-like protein PDGRL platelet derived growth factor receptor like platelet-derived growth factor receptor-like platelet-derived growth factor receptor-like protein platelet-derived growth factor-beta-like tumor suppressor PRLTS; PDGF receptor beta-like tumor suppressor; PDGFR-like protein; Platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor
Gene Aliases: 1110039P19Rik; AV013190; PDGFRL; PDGRL; PRLTS
UniProt ID: (Human) Q15198, (Mouse) Q6PE55, (Rat) Q5RJP7
Entrez Gene ID: (Human) 5157, (Mouse) 68797, (Rat) 290771
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.