Novus Biologicals
Manufacturer Code:NBP2833760.1ML
Catalog # NB031305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human PDGFRL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDGF receptor beta-like tumor suppressor; PDGFR-like protein; PDGFR-like protein PDGRL platelet-derived growth factor receptor-like platelet-derived growth factor receptor-like protein platelet-derived growth factor-beta-like tumor suppressor PRLTSPDGF receptor beta-like tumor suppressor; Platelet-derived growth factor receptor-like protein; platelet-derived growth factor-beta-like tumor suppressor
Gene Aliases: PDGFRL; PDGRL; PRLTS
UniProt ID: (Human) Q15198
Entrez Gene ID: (Human) 5157
Molecular Function:
kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.