Novus Biologicals
Manufacturer Code:NB11060969
Catalog # NB11060969
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0004-8A89 |
Immunogen | N-terminal recombinant human platelet derived growth factor receptor alpha fragment. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha platelet-derived growth factor receptor; Alpha-type platelet-derived growth factor receptor; alpha-type platelet-derived growth factor receptor CD140 antigen-like family member A CD140a CD140a antigen EC 2.7.10 EC 2.7.10.1 MGC74795 PDGFR2 PDGFRA/BCR fusion PDGF-R-alpha platelet-derived growth factor receptor alpha polypeptide rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein RHEPDGFRA; CD140 antigen-like family member A; CD140a; CD140a antigen; PDGF-R-alpha; PDGFR-2; PDGFRA/BCR fusion; Platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein
Gene Aliases: CD140A; GAS9; PDGFR-2; PDGFR2; PDGFRA; RHEPDGFRA
UniProt ID: (Human) Q9UD28
Entrez Gene ID: (Human) 5156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.