Novus Biologicals
Manufacturer Code:NB1205443
Catalog # NB1205443
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phospho peptide containing Tyrosine 572 and 574 of Human PDGF Receptor alpha. This corresponds to Tyrosine 579 and 581 of Human PDGFR beta. The sequence is conserved in PDGFR alpha mouse rat and frog. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha platelet-derived growth factor receptor; Alpha-type platelet-derived growth factor receptor; alpha-type platelet-derived growth factor receptor CD140 antigen-like family member A CD140a CD140a antigen EC 2.7.10 EC 2.7.10.1 MGC74795 PDGFR2 PDGFRA/BCR fusion PDGF-R-alpha platelet-derived growth factor receptor alpha polypeptide rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein RHEPDGFRA; CD140 antigen-like family member A; CD140a; CD140a antigen; PDGF-R-alpha; PDGFR-2; PDGFRA/BCR fusion; Platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; Platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein
Gene Aliases: CD140A; GAS9; PDGFR-2; PDGFR2; PDGFRA; RHEPDGFRA
UniProt ID: (Human) Q9UD28
Entrez Gene ID: (Human) 5156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.