Novus Biologicals
Manufacturer Code:NB11060979G
Catalog # NB11060979G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0013-6D35 |
Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human PDGF-AA recombinant protein. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: PDGF A-chain; PDGF subunit A; PDGF-1; PDGF-1 PDGF1PDGF A-chain PDGF-A Platelet-derived growth factor A chain platelet-derived growth factor alpha chain platelet-derived growth factor alpha isoform 2 preproprotein platelet-derived growth factor alpha polypeptidePDGF subunit A platelet-derived growth factor subunit A; Platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; Platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor subunit A
Gene Aliases: PDGF-A; PDGF1; PDGFA
UniProt ID: (Human) P04085
Entrez Gene ID: (Human) 5154
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.