Novus Biologicals
Manufacturer Code:NBP273305R
Catalog # NBP273305R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 19-247 of human PDGF-D/SCDGFB (NP_079484) produced in E.coli. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: IEGFMSTP036 Iris-expressed growth factor PDGF-D platelet derived growth factor D SCDGF-BMGC26867 SCDGFBplatelet-derived growth factor D spinal cord derived growth factor B Spinal cord-derived growth factor B spinal cord-derived growth factor-B; Iris-expressed growth factor; PDGF-D; PDGFD latent form; PDGFD receptor-binding form; Platelet-derived growth factor D; Platelet-derived growth factor D, latent form; Platelet-derived growth factor D, receptor-binding form; SCDGF-B; spinal cord derived growth factor B; Spinal cord-derived growth factor B; spinal cord-derived growth factor-B
Gene Aliases: IEGF; MSTP036; PDGFD; SCDGF-B; SCDGFB; UNQ1899/PRO4345
UniProt ID: (Human) Q9GZP0
Entrez Gene ID: (Human) 80310
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.