Novus Biologicals
Manufacturer Code:NBP2219560.025MG
Catalog # NB019263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Blocking Assay (BLOCK) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F12 |
Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant PDGF-C. Mouse anti human PDGF-C Q9NRA1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Fallotein; FALLOTEIN PDGF-C platelet derived growth factor C platelet-derived growth factor C SCDGFSpinal cord-derived growth factor secretory growth factor-like protein VEGF-E; PDGF-C; PDGFC latent form; PDGFC receptor-binding form; Platelet-derived growth factor C; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form; SCDGF; secretory growth factor-like protein; Spinal cord-derived growth factor; VEGF-E
Gene Aliases: FALLOTEIN; PDGFC; SCDGF; UNQ174/PRO200
UniProt ID: (Human) Q9NRA1
Entrez Gene ID: (Human) 56034
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.