Novus Biologicals
Manufacturer Code:NBP289373AF647
Catalog # NB088594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human PDGF-C (NP_057289.1 Val235-Gly345). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Fallotein; FALLOTEIN PDGF-C platelet derived growth factor C platelet-derived growth factor C SCDGFSpinal cord-derived growth factor secretory growth factor-like protein VEGF-E; PDGF-C; PDGFC latent form; PDGFC receptor-binding form; Platelet-derived growth factor C; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form; SCDGF; secretory growth factor-like protein; Spinal cord-derived growth factor; VEGF-E
Gene Aliases: FALLOTEIN; PDGFC; SCDGF; UNQ174/PRO200
UniProt ID: (Human) Q9NRA1
Entrez Gene ID: (Human) 56034
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.